High pressure in-line filters
Supplier: VICI JOUR RESEARCH
Certificates
About this item
The stainless steel in-line filter has virtually no dead volume and can be used in various applications without significant band broadening or loss of efficiency. This version is intended for analytical use between the autosampler injection valve and the column.
- Minimises dead volume
- For high pressure applications (up to 350 bar/5000 psi)
Frequently Bought Together