VWR® Hichrom Last Drop™ Inlet Filters
Supplier: Avantor
Certificates
About this item
The Last Drop™ eluent filter utilises a flat element, which sits parallel to the bottom of the reservoir, allowing the filter to draw all but the last 2% of the eluent without drawing air into the system. The filter consists of a 316 stainless steel or PTFE filter element in an inert housing.
- Reduces solvent waste
- Should be used for sensitive biochromatography applications
Delivery information: Every filter is supplied with a universal PEEK tubing adaptor.
Frequently Bought Together