VWR® Hichrom Column End Plugs
: Avantor
These polymer column end plugs are used to prevent solvent evaporation when the columns are not in use.
- Fingertight design
- Extends column life
- Fits any size columns
Frequently Bought Together
These polymer column end plugs are used to prevent solvent evaporation when the columns are not in use.