Back pressure regulator
About this item
This regulator installs on the detector drain line and prevents bubble formation in the flow cell during outgassing. It has an adjustable back pressure from 0 to 20 bar (15 to 300 psi) without disconnecting from the system. Other sizes available.
- Improves baseline stability
- Low dead volume
- Biocompatible
3 Options Available Below
Product Details & Documents
This regulator installs on the detector drain line and prevents bubble formation in the flow cell during outgassing. It has an adjustable back pressure from 0 to 20 bar (15 to 300 psi) without disconnecting from the system. Other sizes available.
- Improves baseline stability
- Low dead volume
- Biocompatible
Product Family Options
Product Information
- PressureAvailabilityPrice
- 554-1492Biocompatible back pressure regulator
- JOURJR-BPR2Biocompatible back pressure regulator
- JOURJR-BPR3Stainless steel back pressure regulator for HPLC/SFCStainless steel back pressure regulator for HPLC/SFC900 - 300 barLog in for availability1 * 1 ST471,00 €