Order Entry
Belgium
ContactUsLinkComponent
Biocompatible mixing tee
Biocompatible mixing tee
Catalog # 554-1547
Supplier:  VICI JOUR RESEARCH
undefined
Biocompatible mixing tee
Catalog # 554-1547
Supplier:  VICI JOUR RESEARCH

Specifications

  • Description:
    Mixing tee with fittings
  • Pk:
    1

Specifications

About this item

PEEK with titanium filter element.

  • Ideal for analytical gradient HPLC with built-in protein friendly PAT filter
  • Totally metal-free
  • Can withstand pressure up to 350 bar
  • For 1/16" OD tubing

Biocompatible. Porosity 10 µm, dead volume 4 µl. Comes complete with three PEEK fingertight fittings.