Orders LinkContactUsLinkComponent
Columns, TSK-Gel® Amide-80
Columns, TSK-Gel® Amide-80
Supplier:   TOSOH Bioscience
Columns, TSK-Gel® Amide-80
Supplier:  TOSOH Bioscience
Certificates
Columns, TSK-Gel® Amide-80
Columns, TSK-Gel® Amide-80
16 Options Available Below
Product Details & Documents
Stainless steel columns with non-ionic carbamoyl [-C(NH₂)O] groups on a silica gel basis suitable for the separation of hydrophilic and polar compounds.
Max. pressure: 150 kg/cm²
Organic solvents: 0–100%
pH: 2 to 7,5
Salt concentration: 100 mM
Temperature stability: 4 - 80 °C
Product Family Options
Guard columns
Product Information
  • Column Format
    Pore size
    Particle Size
    Ø int.
    Length
    Availability
    Price
  • 554-1650
    Amide-80
    Amide-80
    Guard column
    80 Å
    5 µm
    4,60 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 340,00 €
    340,00 €
    1 * 1 ST
       340,00 €
  • Amide-80
    Guard column
    80 Å
    2 µm
    2,00 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 667,00 €
    667,00 €
    1 * 1 ST
       667,00 €
Product Information
  • Column Format
    Pore size
    Particle Size
    Ø int.
    Length
    Availability
    Price
  • Amide-80
    80 Å
    2 µm
    2,00 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 110,00 €
    1 110,00 €
    1 * 1 ST
       1 110,00 €
  • Amide-80
    80 Å
    2 µm
    2,00 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 160,00 €
    1 160,00 €
    1 * 1 ST
       1 160,00 €
  • 554-5942
    Amide-80
    Amide-80
    80 Å
    2 µm
    2,00 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 280,00 €
    1 280,00 €
    1 * 1 ST
       1 280,00 €
  • Amide-80
    80 Å
    2 µm
    3,00 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 110,00 €
    1 110,00 €
    1 * 1 ST
       1 110,00 €
  • Amide-80
    80 Å
    2 µm
    3,00 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 160,00 €
    1 160,00 €
    1 * 1 ST
       1 160,00 €
  • Amide-80
    80 Å
    2 µm
    3,00 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 280,00 €
    1 280,00 €
    1 * 1 ST
       1 280,00 €
  • 554-1601
    Amide-80
    Amide-80
    80 Å
    3 µm
    2,00 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 160,00 €
    1 160,00 €
    1 * 1 ST
       1 160,00 €
  • 554-1651
    Amide-80
    Amide-80
    80 Å
    5 µm
    2,00 mm
    50 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 040,00 €
    1 040,00 €
    1 * 1 ST
       1 040,00 €
  • 554-1652
    Amide-80
    Amide-80
    80 Å
    5 µm
    2,00 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 120,00 €
    1 120,00 €
    1 * 1 ST
       1 120,00 €
  • 554-1414
    Amide-80
    Amide-80
    80 Å
    5 µm
    2,00 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 130,00 €
    1 130,00 €
    1 * 1 ST
       1 130,00 €
  • 554-1415
    Amide-80
    Amide-80
    80 Å
    5 µm
    2,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 150,00 €
    1 150,00 €
    1 * 1 ST
       1 150,00 €
  • 554-1412
    Amide-80
    Amide-80
    80 Å
    5 µm
    4,60 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 220,00 €
    1 220,00 €
    1 * 1 ST
       1 220,00 €
  • 554-1622
    Amide-80
    Amide-80
    80 Å
    10 µm
    7,80 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 240,00 €
    2 240,00 €
    1 * 1 ST
       2 240,00 €
  • 554-1623
    Amide-80
    Amide-80
    80 Å
    10 µm
    21,50 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5 730,00 €
    5 730,00 €
    1 * 1 ST
       5 730,00 €
Accessories
Product Information
  • Catalog #
    Ø int.
    Length
    Availability
    Price
  • Guard cartridge holder for 2 mm I-Ø guard cartridges
    Guard cartridge holder for 2 mm I-Ø guard cartridges
    TOYO0019308
    2,00 mm
    15 mm
    Log in for availability
    1 * 1 ST - 474,00 €
Product Information
  • Catalog #
    Ø int.
    Length
    Availability
    Price
  • Guard cartridge for 2 mm I-Ø columns
    Guard cartridge for 2 mm I-Ø columns
    TOYO0021862
    2,00 mm
    10 mm
    Log in for availability
    1 * 3 ST - 427,00 €