Orders LinkContactUsLinkComponent
Distillation columns, Vigreux
Distillation columns, Vigreux
Distillation columns, Vigreux
Distillation columns, Vigreux
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Air cooled fractionating column with alternating horizontal and inclined indentations.
Product Family Options
  • Usable length
    Cone size
    Socket size
  • 200 mm
    14/23 NS
    14/23 NS
    1 * 1 ST
    1 * 1 ST - 67,00 €
    67,00 €
    1 * 1 ST
       67,00 €
  • 300 mm
    14/23 NS
    14/23 NS
    1 * 1 ST
    1 * 1 ST - 78,50 €
    78,50 €
    1 * 1 ST
       78,50 €
  • 300 mm
    24/29 NS
    24/29 NS
    1 * 1 ST
    1 * 1 ST - 89,80 €
    89,80 €
    1 * 1 ST
       89,80 €
  • 300 mm
    29/32 NS
    29/32 NS
    1 * 1 ST
    1 * 1 ST - 88,60 €
    88,60 €
    1 * 1 ST
       88,60 €
  • 600 mm
    24/29 NS
    24/29 NS
    1 * 1 ST
    1 * 1 ST - 149,00 €
    149,00 €
    1 * 1 ST
       149,00 €
  • 600 mm
    29/32 NS
    29/32 NS
    1 * 1 ST
    1 * 1 ST - 161,00 €
    161,00 €
    1 * 1 ST
       161,00 €