Orders LinkContactUsLinkComponent
Fractionating columns, Dufton, Quickfit®
Fractionating columns, Dufton, Quickfit®
Supplier:   SciLabware
Fractionating columns, Dufton, Quickfit®
Supplier:  SciLabware
Certificates
Fractionating columns, Dufton, Quickfit®
Fractionating columns, Dufton, Quickfit®
2 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass
  • Very good chemical resistance
  • High temperature resistance
Comprises column with accurately fitted Dufton glass spiral. Good efficiency - liquid spiralling down column meets ascending vapour. Spiral can be removed for cleaning.
Product Family Options
Product Information
  • Usable length
    Total length
    Cone size
    Socket size
    Availability
    Price
  • 150 mm
    270 mm
    19/26 NS
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 140,00 €
    140,00 €
    1 * 1 ST
       140,00 €
  • 300 mm
    425 mm
    19/26 NS
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 172,00 €
    172,00 €
    1 * 1 ST
       172,00 €