Orders LinkContactUsLinkComponent
Raschig rings
Raschig rings
Raschig rings
Raschig rings
Soda lime glass.
  • For packing columns
  • Good chemical resistance
  • Good thermal resistance
Product Family Options
  • Description
    Ext.Ø
    Height
  • Raschig ring
    3 mm
    3 mm
    1 * 1 kg
    1 * 1 kg - 209,00 €
    209,00 €
    1 * 1 kg
       209,00 €
  • Raschig ring
    4 mm
    4 mm
    1 * 1 kg
    1 * 1 kg - 155,00 €
    155,00 €
    1 * 1 kg
       155,00 €
  • Raschig ring
    5 mm
    5 mm
    1 * 1 kg
    1 * 1 kg - 138,00 €
    138,00 €
    1 * 1 kg
       138,00 €
  • Raschig ring
    6 mm
    6 mm
    1 * 1 kg
    1 * 1 kg - 87,60 €
    87,60 €
    1 * 1 kg
       87,60 €
  • Raschig ring
    8 mm
    8 mm
    1 * 1 kg
    1 * 1 kg - 86,60 €
    86,60 €
    1 * 1 kg
       86,60 €
  • Raschig ring
    10 mm
    10 mm
    1 * 1 kg
    1 * 1 kg - 83,90 €
    83,90 €
    1 * 1 kg
       83,90 €