Orders LinkContactUsLinkComponent
Overalls, PP
Overalls, PP
White, L
Catalog # 113-0353
Overalls, PP
Catalog # 113-0353
Overalls, PP
Overalls, PP
White, L
Price
64,70 €
Log in for availability
1 * 25 ST
Product Details & Documents
Overalls made from strong lightweight and breathable polypropylene fabric.
  • Protection for various working environment applications
  • Protects against certain hazardous and non hazardous fluids, dry particulates, dirt and grime
Ideal for use in industrial, pharmaceutical, laboratory, clinical, food processing, construction sites, other contaminated areas, and certain environmental cleanup sites.
Weight: 30 g
Colour: White or blue
Specifications
  • Clothing Size:
    L
  • Colour:
    White
  • Seq:
    03
  • Cat. No.:
    113-0353
  • Garment application:
    Laboratory|Clinical|Industrial|Food
  • Weight:
    30 g