Orders LinkContactUsLinkComponent
Jackets, Tyvek® 500, model PP33
Jackets, Tyvek® 500, model PP33
Supplier:   DuPont
Jackets, Tyvek® 500, model PP33
Supplier:  DuPont
Certificates
Jackets, Tyvek® 500, model PP33
4 Options Available Below
Product Details & Documents
These jackets are made from Tyvek®, a flash spun high density PE fabric. Tyvek® is permeable to air and water vapour, but repels aerosols and water-based liquids. It offers an excellent barrier against fine particles and fibres (down to 1 µm in size).
  • Low-linting fabric
  • Antistatic treatment (EN 1149-5) - on both sides
  • With hood and zip closure
  • Stitched seams (internal)
Typical applications include: pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up.
Colour: white
 : (EU) 2016/425; partial body chemical protective clothing, cat III, Type PB [6-B]; EN 14126.
 : 50 per box, bulk packed.
Specifications for Product Family
Specifications
  • Catalog No:
  • 113-3884
  • 113-3885
  • 113-3886
  • 113-3887
  • Model:
  • 500 PP33
  • 500 PP33
  • 500 PP33
  • 500 PP33
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Garment application:
  • Chemical protection|Laboratory|Industrial
  • Chemical protection|Laboratory|Industrial
  • Chemical protection|Laboratory|Industrial
  • Chemical protection|Laboratory|Industrial
Specifications
Product Family Options
Product Information
  • Colour
    Clothing Size
    Availability
    Price
  • White
    M
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 416,00 €
    416,00 €
    1 * 50 ST
       416,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-3884
    Model
    500 PP33
    Gender
    Unisex
    Garment application
    Chemical protection|Laboratory|Industrial
  • White
    L
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 416,00 €
    416,00 €
    1 * 50 ST
       416,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-3885
    Model
    500 PP33
    Gender
    Unisex
    Garment application
    Chemical protection|Laboratory|Industrial
  • White
    XL
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 416,00 €
    416,00 €
    1 * 50 ST
       416,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-3886
    Model
    500 PP33
    Gender
    Unisex
    Garment application
    Chemical protection|Laboratory|Industrial
  • White
    XXL
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 416,00 €
    416,00 €
    1 * 50 ST
       416,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-3887
    Model
    500 PP33
    Gender
    Unisex
    Garment application
    Chemical protection|Laboratory|Industrial