Orders LinkContactUsLinkComponent
Beard covers, PP
Beard covers, PP
With latex, White
Catalog # 113-0240
Beard covers, PP
Catalog # 113-0240
Beard covers, PP
Beard covers, PP
With latex, White
Price
69,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
This beard restraint made from PP provides economical protection for various working environment applications. It protects workers against certain hazardous and non hazardous fluids, dry particulates, dirt and grime.
  • Lightweight
  • Breathable fabric
Latex-free version available: 113-0352.
Specifications
  • Colour:
    White
  • Description:
    With latex