Drip closures
Supplier: KARTELL
Jump to:
About this item
HDPE, natural colour.
- Good chemical resistance, good temperature resistance
- Available in two lengths
- Attached cap
For narrow-neck bottles with round thread in accordance with DIN 168.
5 Options Available Below
Product Details & Documents
HDPE, natural colour.
- Good chemical resistance, good temperature resistance
- Available in two lengths
- Attached cap
For narrow-neck bottles with round thread in accordance with DIN 168.