Bottles, narrow neck, with thread
About this item
Glass, clear, amber or yellow.
- Neck Ø: 28 mm
- Good chemical and temperature resistance
- Amber coloured to protect light sensitive substances
Choice of various screw caps.
Ordering information: Supplied without screw caps - please order separately.
Packaging: Packaged in impermeable plastic packs.
23 Options Available Below
Product Details & Documents
Glass, clear, amber or yellow.
- Neck Ø: 28 mm
- Good chemical and temperature resistance
- Amber coloured to protect light sensitive substances
Choice of various screw caps.
Ordering information: Supplied without screw caps - please order separately.
Packaging: Packaged in impermeable plastic packs.
Documents
Product Family Options
White glass bottles, narrow neck Ø 28 mm, without screw cap
Product Information
- CapacityAvailabilityPrice
Amber glass bottles, narrow neck Ø 28 mm, without screw cap
Product Information
- CapacityAvailabilityPrice
Yellow glass bottles, narrow neck ∅ 28 mm, without screw cap
Product Information
- CapacityAvailabilityPrice
Bakelite screw caps with seal
Product Information
- Catalog #ColourThreadDescriptionAvailabilityPrice
Aluminium screw caps with seal
Product Information
- Catalog #ColourDescriptionThreadAvailabilityPrice
Polypropylene screw caps
Product Information
- Catalog #ColourDescriptionThreadAvailabilityPrice