Screw caps for square bottles, DURAN®
PP, red or blue.
- Standard caps, blue, with snap ring for narrow mouth and wide mouth bottles
- Safety caps with valve (valve cap opens with 0,1 bar overpressure), for narrow mouth bottles
- Very good chemical and temperature resistance, leakproof
Tamper evident caps for narrow mouth and wide mouth square bottles with DIN thread.
PP, red or blue.
- Standard caps, blue, with snap ring for narrow mouth and wide mouth bottles
- Safety caps with valve (valve cap opens with 0,1 bar overpressure), for narrow mouth bottles
- Very good chemical and temperature resistance, leakproof
Tamper evident caps for narrow mouth and wide mouth square bottles with DIN thread.
Product Family Options
- DescriptionThreadColour
- Standard screw caps for narrow mouth bottlesGL 32 HBlue1 * 10 ST18,60 €
- Safety screw caps for narrow mouth bottlesGL 32 HRed1 * 10 ST61,00 €
- Standard screw caps for wide mouth bottlesGL 32 KBlue1 * 10 ST13,00 €
- Standard screw caps for narrow mouth bottlesGL 45 HBlue1 * 10 ST20,10 €
- Standard screw caps for wide mouth bottlesGL 45 KBlue1 * 10 ST18,60 €
- Standard screw caps for wide mouth bottlesGL 54 KBlue1 * 10 ST21,50 €
- Standard screw caps for wide mouth bottlesGL 60 KBlue1 * 10 ST18,80 €