Orders LinkContactUsLinkComponent
Nalgene™ Screw Caps
   Sustainable Options Available
Nalgene™ Screw Caps
   Sustainable Options Available
Certificates
Nalgene™ Screw Caps
52 Options Available Below
Product Details & Documents
PP, coloured.
  • Very good chemical resistance
  • Heat resistant up to 135 °C
  • Autoclavable
Screw caps without inserts for Nalgene® narrow and wide mouth bottles, as well as Erlenmeyer flasks with thread.
Specifications for Product Family
Specifications
  • Catalog No:
  • 215-1591
  • 215-1597
  • 215-1598
  • 215-1236
  • 215-1247
  • 215-1599
  • 215-1248
  • 215-1249
  • 215-1600
  • 215-1601
  • 215-1602
  • 215-1250
  • 215-1603
  • 215-1604
  • 215-1605
  • 215-1606
  • 215-1251
  • 215-1607
  • 215-1608
  • 215-1609
  • 215-1610
  • 215-1252
  • 215-1611
  • 215-1612
  • 215-1613
  • 215-1618
  • 215-1619
  • 215-1620
  • 215-1621
  • 215-1622
  • 216-8953
  • 216-8954
  • 216-8957
  • 216-8959
  • 216-8960
  • 216-8961
  • NALG71-2151-0053
  • NALG71-2151-0083
  • NALG71-2151-0200
  • NALG71-2151-0380
  • NALG71-2154-0700
  • NALG712155-1200
  • NALG71-2160-0384
  • 216-8972
  • 216-8973
  • NALG71-2171-0530
  • NALG71-2172-0020
  • NALG712174-0530
  • NALG71-2174-0530
  • NALG71-2240-1053
  • NALG71-2411-1030
  • NALG71-2411-1125
  • NALG71-2401-0001
  • NALG71-2401-0002
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
  • Cap Size
    Colour
    Material
    Availability
    Price
  • 215-1591
    Linerless
    Linerless
    13-415
    Natural
    PP
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 664,00 €
    664,00 €
    1 * 2.000 ST
       664,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1591
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1597
    Linerless
    Linerless
    20-415
    Natural
    PP
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 725,00 €
    725,00 €
    1 * 2.000 ST
       725,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1597
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1598
    Linerless
    Linerless
    24-415
    Natural
    PP
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 452,00 €
    452,00 €
    1 * 1.000 ST
       452,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1598
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1236
    Linerless
    Linerless
    28-415
    Natural
    PP
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 283,00 €
    283,00 €
    1 * 500 ST
       283,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1236
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Linerless
    38-430
    Natural
    PP
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 177,00 €
    177,00 €
    1 * 250 ST
       177,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1247
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1599
    Linerless
    Linerless
    13-415
    White
    PP
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 713,00 €
    713,00 €
    1 * 2.000 ST
       713,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1599
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1248
    Linerless
    Linerless
    20-415
    White
    PP
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 542,00 €
    542,00 €
    1 * 2.000 ST
       542,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1248
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1249
    Linerless
    Linerless
    24-415
    White
    PP
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 350,00 €
    350,00 €
    1 * 1.000 ST
       350,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1249
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1600
    Linerless
    Linerless
    28-415
    White
    PP
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 297,00 €
    297,00 €
    1 * 500 ST
       297,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1600
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1601
    Linerless
    Linerless
    38-430
    White
    PP
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 194,00 €
    194,00 €
    1 * 250 ST
       194,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1601
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1602
    Linerless
    Linerless
    13-415
    Green
    PP
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 713,00 €
    713,00 €
    1 * 2.000 ST
       713,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1602
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1250
    Linerless
    Linerless
    20-415
    Green
    PP
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 770,00 €
    770,00 €
    1 * 2.000 ST
       770,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1250
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1603
    Linerless
    Linerless
    24-415
    Green
    PP
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 497,00 €
    497,00 €
    1 * 1.000 ST
       497,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1603
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1604
    Linerless
    Linerless
    28-415
    Green
    PP
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 297,00 €
    297,00 €
    1 * 500 ST
       297,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1604
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1605
    Linerless
    Linerless
    38-430
    Green
    PP
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 194,00 €
    194,00 €
    1 * 250 ST
       194,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1605
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1606
    Linerless
    Linerless
    13-415
    Red
    PP
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 713,00 €
    713,00 €
    1 * 2.000 ST
       713,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1606
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1251
    Linerless
    Linerless
    20-415
    Red
    PP
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 542,00 €
    542,00 €
    1 * 2.000 ST
       542,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1251
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1607
    Linerless
    Linerless
    24-415
    Red
    PP
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 497,00 €
    497,00 €
    1 * 1.000 ST
       497,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1607
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1608
    Linerless
    Linerless
    28-415
    Red
    PP
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 297,00 €
    297,00 €
    1 * 500 ST
       297,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1608
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1609
    Linerless
    Linerless
    38-430
    Red
    PP
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 194,00 €
    194,00 €
    1 * 250 ST
       194,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1609
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1610
    Linerless
    Linerless
    13-415
    Blue
    PP
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 713,00 €
    713,00 €
    1 * 2.000 ST
       713,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1610
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1252
    Linerless
    Linerless
    20-415
    Blue
    PP
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 770,00 €
    770,00 €
    1 * 2.000 ST
       770,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1252
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1611
    Linerless
    Linerless
    24-415
    Blue
    PP
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 497,00 €
    497,00 €
    1 * 1.000 ST
       497,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1611
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1612
    Linerless
    Linerless
    28-415
    Blue
    PP
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 297,00 €
    297,00 €
    1 * 500 ST
       297,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1612
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1613
    Linerless
    Linerless
    38-430
    Blue
    PP
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 194,00 €
    194,00 €
    1 * 250 ST
       194,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1613
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1618
    Linerless
    Linerless
    13-415
    Amber
    PP
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 722,00 €
    722,00 €
    1 * 2.000 ST
       722,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1618
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1619
    Linerless
    Linerless
    20-415
    Amber
    PP
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 779,00 €
    779,00 €
    1 * 2.000 ST
       779,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1619
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1620
    Linerless
    Linerless
    24-415
    Amber
    PP
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 502,00 €
    502,00 €
    1 * 1.000 ST
       502,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1620
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1621
    Linerless
    Linerless
    28-415
    Brown
    PP
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 300,00 €
    300,00 €
    1 * 500 ST
       300,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1621
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 215-1622
    Linerless
    Linerless
    38-430
    Amber
    PP
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 195,00 €
    195,00 €
    1 * 250 ST
       195,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    215-1622
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 216-8953
    Replacement closure
    Replacement closure
    24 mm
    Natural
    PP
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 14,80 €
    14,80 €
    1 * 12 ST
       14,80 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-8953
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 216-8954
    Replacement closure
    Replacement closure
    28 mm
    Natural
    PP
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 15,70 €
    15,70 €
    1 * 12 ST
       15,70 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-8954
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 216-8957
    Replacement closure
    Replacement closure
    43 mm
    Natural
    PP
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 18,70 €
    18,70 €
    1 * 12 ST
       18,70 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-8957
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 216-8959
    Replacement closure
    Replacement closure
    53 mm
    Natural
    PP
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 27,20 €
    27,20 €
    1 * 12 ST
       27,20 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-8959
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 216-8960
    Replacement closure
    Replacement closure
    63 mm
    Natural
    PP
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 39,40 €
    39,40 €
    1 * 12 ST
       39,40 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-8960
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 216-8961
    Replacement closure
    Replacement closure
    100 mm
    White
    PP
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 62,40 €
    62,40 €
    1 * 12 ST
       62,40 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-8961
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • NALG71-2151-0053
    Replacement closure
    Replacement closure
    53B
    White
    PP
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 93,80 €
    93,80 €
    1 * 12 ST
       93,80 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG71-2151-0053
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • NALG71-2151-0083
    Replacement closure
    Replacement closure
    83 B
    Natural
    PP
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 60,00 €
    60,00 €
    1 * 2 ST
       60,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG71-2151-0083
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • NALG71-2151-0200
    Replacement closure
    Replacement closure
    20 mm
    Clear
    PP
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 14,00 €
    14,00 €
    1 * 12 ST
       14,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG71-2151-0200
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • NALG71-2151-0380
    Replacement closure
    Replacement closure
    38 mm
    Natural
    PP
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 17,90 €
    17,90 €
    1 * 12 ST
       17,90 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG71-2151-0380
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • NALG71-2154-0700
    Replacement closure
    Replacement closure
    70 mm
    Natural
    PP
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 39,50 €
    39,50 €
    1 * 12 ST
       39,50 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG71-2154-0700
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • NALG712155-1200
    Replacement closure
    Replacement closure
    120 mm
    White
    PP
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    NALG712155-1200
    Environmentally Preferable
    Environmental certification
  • Replacement closure
    38-430 mm
    Natural
    PP
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 21,30 €
    21,30 €
    1 * 12 ST
       21,30 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG71-2160-0384
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 216-8972
    Replacement closure
    Replacement closure
    53B
    White
    PP
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 134,00 €
    134,00 €
    1 * 12 ST
       134,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-8972
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 216-8973
    Replacement closure
    Replacement closure
    83B
    White
    PP
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 60,00 €
    60,00 €
    1 * 2 ST
       60,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    216-8973
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • NALG71-2171-0530
    Replacement closure
    Replacement closure
    53 B
    Amber
    PP
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 31,10 €
    31,10 €
    1 * 12 ST
       31,10 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG71-2171-0530
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • NALG71-2172-0020
    Replacement closure
    Replacement closure
    20 mm
    Natural
    PP
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 34,10 €
    34,10 €
    1 * 2 ST
       34,10 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG71-2172-0020
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • NALG712174-0530
    Replacement closure
    Replacement closure
    53 mm
    Natural
    PP
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    NALG712174-0530
    Environmentally Preferable
    Environmental certification
  • NALG71-2174-0530
    Replacement closure
    Replacement closure
    53 mm
    Clear
    PP
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 56,00 €
    56,00 €
    1 * 2 ST
       56,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    NALG71-2174-0530
    Environmentally Preferable
    Environmental certification
  • NALG71-2240-1053
    Replacement closure
    Replacement closure
    53 mm
    PP
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 286,00 €
    286,00 €
    1 * 10 ST
       286,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG71-2240-1053
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • NALG71-2411-1030
    Replacement closure
    Replacement closure
    20 mm
    PP
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 150,00 €
    150,00 €
    1 * 25 ST
       150,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG71-2411-1030
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • NALG71-2411-1125
    Replacement closure
    Replacement closure
    24 mm
    PP
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 155,00 €
    155,00 €
    1 * 25 ST
       155,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG71-2411-1125
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • NALG71-2401-0001
    With stem and draw tube for wash bottle
    With stem and draw tube for wash bottle
    28 mm
    Natural
    PP
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 167,00 €
    167,00 €
    1 * 25 ST
       167,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG71-2401-0001
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • NALG71-2401-0002
    With stem and draw tube for wash bottle
    With stem and draw tube for wash bottle
    24 mm
    Natural
    PP
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 168,00 €
    168,00 €
    1 * 25 ST
       168,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    NALG71-2401-0002
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
Replacement gaskets
Product Information
  • Catalog #
    For
    Description
    Availability
    Price
  • 53B
    Sealing gaskets
    Log in for availability
    Log in for availability
    1 * 12 ST
    1 * 12 ST - 34,70 €
    34,70 €
    1 * 12 ST
       34,70 €
Product Information
  • Catalog #
    For
    Description
    Availability
    Price
  • 83B
    Sealing gaskets
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 62,80 €
    62,80 €
    1 * 5 ST
       62,80 €