Forceps, Nalgene®
229-0301
- Colour:Blue
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:NAR Certificate of Retirement
- Pk:12
PP, blue. With double-action hinges for extra leverage. The jaws have interlocking teeth for holding objects securely. Scissor type handles with catch for locking the forceps. Can be used as clips for chromatography columns. Also serve as excellent tube clamps.
- Good chemical resistance
- Very good thermal resistance, autoclavable
- Convenient and easy to use, durable