Orders LinkContactUsLinkComponent
Clamps
Clamps
Supplier:   USBECK Laborgeräte
Clamps
Certificates
Clamps
4 Options Available Below
Product Details & Documents
Zinc die-casting, nickel plated.
  • With vinyl coated jaws
  • With rod Ø 10 mm
Product Family Options
Product Information
  • Spanning width
    Availability
    Price
  • 30 - 80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,30 €
    23,30 €
    1 * 1 ST
       23,30 €
  • 12 - 25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,40 €
    18,40 €
    1 * 1 ST
       18,40 €
  • 20 - 40 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,70 €
    19,70 €
    1 * 1 ST
       19,70 €
  • 38 - 60 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 21,00 €
    21,00 €
    1 * 1 ST
       21,00 €