Orders LinkContactUsLinkComponent
Clamps, universal
Clamps, universal
Supplier:   USBECK Laborgeräte
Clamps, universal
Certificates
Clamps, universal
Clamps, universal
4 Options Available Below
Product Details & Documents
The fingers of the clamps have cork inserts to protect the clamped items.
  • Without bosshead for fixing to stand
  • With T-screws for rapid easy clamping
Product Family Options
Product Information
  • Material
    Spanning width
    Availability
    Price
  • Zinc die-cast, powder coated
    0 - 80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 17,80 €
    17,80 €
    1 * 1 ST
       17,80 €
  • Stainless steel
    0 - 80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 74,40 €
    74,40 €
    1 * 1 ST
       74,40 €
  • Nickel coated steel
    0 - 80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,90 €
    23,90 €
    1 * 1 ST
       23,90 €
  • Zinc die-cast, natural alloy
    0 - 80 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16,40 €
    16,40 €
    1 * 1 ST
       16,40 €