Orders LinkContactUsLinkComponent
Inoculating loops, reusable
Inoculating loops, reusable
Supplier:   RSG
Inoculating loops, reusable
Supplier:  RSG
Certificates
Inoculating loops, reusable
Inoculating loops, reusable
6 Options Available Below
Product Details & Documents
These NiCr 8020 loops are available in a range of sizes and lengths with internal diameter of 1,5, 2,5 or 4 mm. They are heat resistant and can be flame sterilised. Diameter of wire: 0,6 mm. Loop holder available separately.
Documents
Product Family Options
Product Information
  • Availability
    Price
  • 631-7131
    Loops, length 50 mm, Ø 1,5 mm
    Loops, length 50 mm, Ø 1,5 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 13,40 €
    13,40 €
    1 * 10 ST
       13,40 €
  • 631-7133
    Loops, length 50 mm, Ø 4,0 mm
    Loops, length 50 mm, Ø 4,0 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 13,40 €
    13,40 €
    1 * 10 ST
       13,40 €
  • 631-7132
    Loops, length 50 mm, Ø 2,5 mm
    Loops, length 50 mm, Ø 2,5 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 28,20 €
    28,20 €
    1 * 10 ST
       28,20 €
  • 631-0086
    Loops, length 75 mm, Ø 4,0 mm
    Loops, length 75 mm, Ø 4,0 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 15,40 €
    15,40 €
    1 * 10 ST
       15,40 €
  • 631-0085
    Loops, length 75 mm, Ø 2,5 mm
    Loops, length 75 mm, Ø 2,5 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 15,40 €
    15,40 €
    1 * 10 ST
       15,40 €
  • 631-0084
    Loops, length 75 mm, Ø 1,5 mm
    Loops, length 75 mm, Ø 1,5 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 15,40 €
    15,40 €
    1 * 10 ST
       15,40 €
Accessories
Product Information
  • Catalog #
    Length
    Availability
    Price
  • Needle holder
    Needle holder
    391-0374
    240 mm
    Log in for availability
    1 * 1 ST - 15,00 €
Product Information
  • Catalog #
    Length
    Availability
    Price
  • Needle holder
    Needle holder
    391-0373
    170 mm
    Log in for availability
    1 * 1 ST - 13,00 €