VWR®, Inoculating Loops and Needles, Disposable
Catalog # 612-9358
Supplier: VWR Collection
Specifications
- Packed:20/bag
- Capacity:10 µl
- Colour:Light blue
- Description:Loops, flexible
- Pk:1.000
Specifications
About this item
PS, sterile. These loops ensure consistent wetting and complete liquid transfer. Available rigid or flexible and have smooth edges to avoid gouging the surface of the media.
- Gamma sterilised
- Colour coded
- Hexagonal shaft for easy handling
Delivery information: Supplied with a calibration certificate in each box.