Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Inoculating loops, Microstreaker
Inoculating loops, Microstreaker
Certificates
Inoculating loops, Microstreaker
Inoculating loops, Microstreaker
3 Options Available Below
Product Details & Documents
Inoculation loop of nickel-chromium wire of not more than 60 mm length, conforming to Howie recommendations. Colour coded handle.
Product Family Options
Product Information
  • Désignation
    Capacité
    Availability
    Price
  • Petit, diamètre int. de 2,26 mm, poignée bleue
    2 µl
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 99,10 €
    99,10 €
    1 * 5 ST
       99,10 €
  • Moyen, diamètre int. de 2,91 mm, poignée verte
    3 µl
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 102,00 €
    102,00 €
    1 * 5 ST
       102,00 €
  • Grand, diamètre int. de 5,05 mm, poignée rouge
    10 µl
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 89,80 €
    89,80 €
    1 * 5 ST
       89,80 €