ContactUsLinkComponent
Safety bottle carrier with lid, Nalgene®
Safety bottle carrier with lid, Nalgene®
Catalog # 216-4428
   Low Manufacturing Impact
undefined
Safety bottle carrier with lid, Nalgene®
Catalog # 216-4428
   Low Manufacturing Impact
Specifications
  • For:
    6×500 ml bottles
  • Colour:
    Yellow
  • Description:
    Safety transport basket
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    1
Specifications
About this item
To hold and carry up to six 500 ml standard reagent bottles, and most other glass or plastic 500 ml bottles, this carrier consists of a bottle basket and a tight fitting cover.
LDPE.
  • Stackable
  • Holds bottles securely
  • Minimise risk of bottles smashing together, falling out or breaking
  • No splashing of dangerous chemicals if glass bottles break
  • Sturdy, wide grip for easy carrying