Orders LinkContactUsLinkComponent
Burette self-zero kit, Nalgene®
Self-Zero Kit
  612-2587
   Low Manufacturing Impact
Burette self-zero kit, Nalgene®
  612-2587
   Low Manufacturing Impact
Burette self-zero kit, Nalgene®
Burette self-zero kit, Nalgene®
Self-Zero Kit
49,70 €
1 * 1 ST
Makes any plastic or glass burette self-zeroing.
Consists of a 1000 ml LDPE bottle, 1,2 m clear PVC tubing, PP fill tube and adapter.
  • Description:
    Self-Zero Kit
  • Cat. No.:
    612-2587
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact