Orders LinkContactUsLinkComponent
Burettes, Nalgene®
   Sustainable Options Available
Burettes, Nalgene®
   Sustainable Options Available
Certificates
Burettes, Nalgene®
2 Options Available Below
Product Details & Documents
Crystal-clear acrylic, PMP tip and TFE stopcock and plug.
  • Unaffected by dilute mineral acids and bases except HF and NH₄OH
  • Not designed for alcohol and organic solvents
Durable, excellent for general use, schools, industrial labs and field use.
Certifications: ASTM E287
Documents
Specifications for Product Family
Specifications
  • Catalog No:
  • 612-2558
  • 612-2559
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
  • Capacity
    Division
    Tolerance
    Availability
    Price
  • 25 ml
    0,1 ml
    0,06 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 193,00 €
    193,00 €
    1 * 1 ST
       193,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    612-2558
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 50 ml
    0,1 ml
    0,1 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 223,00 €
    223,00 €
    1 * 1 ST
       223,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    612-2559
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Stopcock assembly for burette, 25 ml
    Stopcock assembly for burette, 25 ml
    NALG71-3650-1025
    Log in for availability
    1 * 2 ST - 377,00 €