Order Entry
Belgium
Orders LinkContactUsLinkComponent
Dafert burettes, Schellbach

Dafert burettes, Schellbach

50 ml, 0,1 ml, 0,075 ± ml
Catalog # 612-3589
Supplier:   HIRSCHMANN
Dafert burettes, Schellbach
Catalog # 612-3589
Supplier:  HIRSCHMANN
Dafert burettes, Schellbach
Dafert burettes, Schellbach

50 ml, 0,1 ml, 0,075 ± ml

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

DURAN®, borosilicate glass, class B.

  • Calibrated to deliver 'Ex'
  • Shellbach stripe, short marks at major graduations, blue graduations
  • Stopcock with glass key with two oblique bores for convenient filling and titrating
Specifications
  • Capacity:
    50 ml
  • Seq:
    02
  • Division:
    0,1 ml
  • Tolerance:
    0,075 ± ml