Orders LinkContactUsLinkComponent
Burettes, pellet pattern, lateral and intermediate stopcocks, Schellbach
Burettes, pellet pattern, lateral and intermediate stopcocks, Schellbach
Supplier:   HIRSCHMANN
Burettes, pellet pattern, lateral and intermediate stopcocks, Schellbach
Supplier:  HIRSCHMANN
Certificates
Burettes,  pellet pattern, lateral and intermediate stopcocks, Schellbach
Burettes, pellet pattern, lateral and intermediate stopcocks, Schellbach
5 Options Available Below
Product Details & Documents
DURAN®, class AS.
  • Calibrated to deliver 'Ex', conformity certified (DE-M marking) with dated batch identification
  • Schellbach stripe, ring marks at major graduations, blue graduations
  • Automatic zeroing, glass spindle and with intermediate stopcock with glass key or PTFE spindle
DIN EN ISO 385
Certifications: DIN EN ISO 385
Product Family Options
Lateral and intermediate stopcocks with glass key without reservoir 2 litre bottle
Product Information
  • Capacity
    Division
    Tolerance
    Availability
    Price
  • 10 ml
    0,02 ml
    0,02 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 299,00 €
    299,00 €
    1 * 1 ST
       299,00 €
  • 25 ml
    0,05 ml
    0,03 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 299,00 €
    299,00 €
    1 * 1 ST
       299,00 €
  • 50 ml
    0,1 ml
    0,05 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 299,00 €
    299,00 €
    1 * 1 ST
       299,00 €
Lateral and intermediate stopcocks with PTFE key without reservoir 2 litre bottle
Product Information
  • Capacity
    Division
    Tolerance
    Availability
    Price
  • 25 ml
    0,05 ml
    0,03 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 367,00 €
    367,00 €
    1 * 1 ST
       367,00 €
  • 50 ml
    0,1 ml
    0,05 ± ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 367,00 €
    367,00 €
    1 * 1 ST
       367,00 €
Reservoir bottle
Product Information
  • Catalog #
    Description
    Capacity
    Availability
    Price
  • Reservoir bottle, NS 29/32, DURAN® clear glass
    2000 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31,20 €
    31,20 €
    1 * 1 ST
       31,20 €