Automatic burettes, Dr. Schilling pattern, Schellbach
Supplier: HIRSCHMANN
Jump to:
About this item
Borosilicate 3.3 glass, silicone tubing, PVC fitting, PE bottle and PP base.
- Schellbach stripe, blue graduations
- Automatic zeroing, squeezing bottle allows rapid filling, micrometre screw allows fine titration
With bottle: 500 ml for 5 and 10 ml; 1000 ml for 15, 25 and 50 ml.
7 Options Available Below
Product Details & Documents
Borosilicate 3.3 glass, silicone tubing, PVC fitting, PE bottle and PP base.
- Schellbach stripe, blue graduations
- Automatic zeroing, squeezing bottle allows rapid filling, micrometre screw allows fine titration
With bottle: 500 ml for 5 and 10 ml; 1000 ml for 15, 25 and 50 ml.
Documents
Product Family Options
Product Information
- CapacityDivisionScale lengthToleranceAvailabilityPrice
Bases
Product Information
Accessories
Product Information
- Catalog #CapacityDivisionToleranceAvailabilityPrice
Product Information
- Catalog #CapacityDivisionToleranceAvailabilityPrice
Product Information
- Catalog #CapacityDivisionToleranceAvailabilityPrice
Product Information
- Catalog #CapacityDivisionToleranceAvailabilityPrice
Product Information
- Catalog #CapacityDivisionToleranceAvailabilityPrice
Product Information
- Catalog #AvailabilityPrice
- Fittings for Dr Schilling burette 5 and 10 mlFittings for Dr Schilling burette 5 and 10 mlHIRS8900710Log in for availability1 * 1 ST - 23,70 €
Product Information
- Catalog #AvailabilityPrice
- Fittings for Dr Schilling burette 15-50 ml
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.