Orders LinkContactUsLinkComponent
Buckets
Buckets
Certificates
Buckets
3 Options Available Below
Product Details & Documents
Stainless steel, 304 grade.
  • Very good chemical resistance, very good thermal resistance
  • High quality, very stable
  • An optional lid is available which is extended to cover the spout
These buckets are polished throughout, graduated in 1 litre intervals and come complete with swing handle. Skirtbands are standard. They have crevice and pit free interiors and are polished throughout.
Product Family Options
Product Information
  • Capacity
    Ø top/Ø bottom
    Height
    Availability
    Price
  • 10 L
    295/170 mm
    270 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 184,00 €
    184,00 €
    1 * 1 ST
       184,00 €
  • 12,5 L
    320/210 mm
    260 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 200,00 €
    200,00 €
    1 * 1 ST
       200,00 €
  • 15 L
    340/210 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 216,00 €
    216,00 €
    1 * 1 ST
       216,00 €