Orders LinkContactUsLinkComponent
Buckets
Buckets
Supplier:   KARTELL
Buckets
Supplier:  KARTELL
Certificates
Buckets
3 Options Available Below
Product Details & Documents
LDPE, white.
  • Good chemical and temperature resistance
  • Graduated with litre scale
  • With spout
For industrial and laboratory applications. With steel bracket and plastic grip.
Product Family Options
Product Information
  • Colour
    Capacity
    Ø ext.
    Height
    Availability
    Price
  • White
    9 L
    280 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 21,80 €
    21,80 €
    1 * 1 ST
       21,80 €
  • White
    12 L
    300 mm
    280 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 27,70 €
    27,70 €
    1 * 1 ST
       27,70 €
  • White
    17 L
    340 mm
    310 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 39,30 €
    39,30 €
    1 * 1 ST
       39,30 €