ContactUsLinkComponent
Buckets, round, Nalgene®
Buckets, round, Nalgene®
  216-0093
   Low Manufacturing Impact
undefined
Buckets, round, Nalgene®
  216-0093
   Low Manufacturing Impact
  • Capacity:
    9,5 L
  • Colour:
    White
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Pk:
    1
HDPE, white with spout.
  • Tough and durable
  • Graduated in pins and litres
  • Fitted with polethylene coated wire handle
Ideal for transfer and transport of laboratory solutions and powders.