0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Brushes
Brush, double, for cups and beakers
Brush, double, for cups and beakers
HECH91120000013
:
GLASWARENFABRIK KARL HECHT
Brush, double, for cups and beakers
HECH91120000013
:
GLASWARENFABRIK KARL HECHT
Brush head Ø:
80 mm
Total length:
430 mm
Brush head length:
130 mm
Pk:
50