Orders LinkContactUsLinkComponent
Assistent® Pipette brushes
Assistent® Pipette brushes
Assistent® Pipette brushes
Certificates
Assistent® Pipette brushes
Assistent® Pipette brushes
8 Options Available Below
Product Details & Documents
Straight brushes in different sizes.
Documents
Product Family Options
Product Information
  • Brush head length
    Brush head Ø
    Total length
    Availability
    Price
  • 65 mm
    11 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 171,00 €
    171,00 €
    1 * 50 ST
       171,00 €
  • 70 mm
    8 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 107,00 €
    107,00 €
    1 * 50 ST
       107,00 €
  • 70 mm
    15 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 225,00 €
    225,00 €
    1 * 50 ST
       225,00 €
  • 80 mm
    4 mm
    400 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 306,00 €
    306,00 €
    1 * 50 ST
       306,00 €
  • 90 mm
    30 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 238,00 €
    238,00 €
    1 * 50 ST
       238,00 €
  • 90 mm
    3 mm
    400 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 188,00 €
    188,00 €
    1 * 50 ST
       188,00 €
  • 110 mm
    8 mm
    500 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 518,00 €
    518,00 €
    1 * 50 ST
       518,00 €
  • 110 mm
    8 mm
    850 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 164,00 €
    164,00 €
    1 * 10 ST
       164,00 €