Order Entry
Belgium
Orders LinkContactUsLinkComponent
Storage Boxes

Storage Boxes

4000 ml, 340×230×94 mm
Catalog # 216-3192
Supplier:   VITLAB
Storage Boxes
Catalog # 216-3192
Supplier:  VITLAB
Storage Boxes
Storage Boxes

4000 ml, 340×230×94 mm

Price
40,20 €
Log in for availability
1 * 1 ST
Product Details & Documents

SAN, transparent.
Rectangular, multipurpose container.

  • With lid
  • Good chemical resistance
  • Light and stackable
Specifications
  • Capacity:
    4000 ml
  • W×D×H:
    340×230×94 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.