Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Storage Boxes

Supplier:   Gilac PROFESSIONNEL
Storage Boxes
Certificates
Storage Boxes

17 Options Available Below

Product Details & Documents

PP, coloured.
Ideal for a wide range of uses. With tightly fitting lid.

  • Very good chemical resistance
  • Very good thermal resistance
  • Very sturdy and economical
Product Family Options

Product Information

  • Colour
    W×D×H
    Availability
    Price
  • Red
    395×295×160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 56,90 €
    56,90 €
    1 * 1 ST
       56,90 €

  • Blue
    395×295×160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 56,90 €
    56,90 €
    1 * 1 ST
       56,90 €

  • Green
    395×295×160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 56,90 €
    56,90 €
    1 * 1 ST
       56,90 €

  • Yellow
    400×300×180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • Red
    395×295×215 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • Blue
    395×295×215 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • Green
    395×295×215 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • Blue
    595×395×165 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • Red
    600×400×190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • Green
    600×400×190 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • Red
    595×395×215 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • Blue
    595×395×215 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 106,00 €
    106,00 €
    1 * 1 ST
       106,00 €

  • Green
    595×395×215 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • Red
    595×395×320 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • Green
    595×395×320 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • Blue
    595×395×320 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • White
    600×400×356 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.