Order Entry
Belgium
Orders LinkContactUsLinkComponent
Cryoboxes, PS

Cryoboxes, PS

64, 0,5/1,5/2,0 ml, Blue/white
Catalog # 720-0503
Supplier:   Simport Scientific
Cryoboxes, PS
Catalog # 720-0503
Supplier:  Simport Scientific
Cryoboxes, PS
Cryoboxes, PS

64, 0,5/1,5/2,0 ml, Blue/white

Price
56,20 €
Log in for availability
1 * 4 ST
Product Details & Documents

PS, white/transparent with blue polyurethane foam insert.

  • Boxes have a usable range of –90 to +80 °C
  • Boxes are stackable for space saving in freezers, refrigerators and lab counters
  • Being printed with a series of squares (numbered 1 to 64)

Ideal for holding up to 64 tubes from 0,5 to 2 ml. A transparent cover allows the user to see the contents and is aligned in a specific orientation. The surface accepts writing with markers for better inventory control.

Specifications
  • Compartments:
    64
  • For:
    0,5/1,5/2,0 ml
  • Colour:
    Blue/white
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.