Orders LinkContactUsLinkComponent
Slide boxes, SlideFile™
Slide boxes, SlideFile™
Supplier:   Simport Scientific
Slide boxes, SlideFile™
Supplier:  Simport Scientific
Certificates
Slide boxes, SlideFile™
Slide boxes, SlideFile™
5 Options Available Below
Product Details & Documents
This impact resistant storage system can hold up to 400 slides per unit, for space efficient storage. All slides are stored upright for easier manipulation.
  • Removable tray inside the storage box for 100 individual numbered slots
  • Transparent hinged cover makes it easy to see the contents of the box at a glance
  • Unique feature with this system to read barcodes without having to remove the slides from the box
W×D×H: 82×245×86 mm
Ordering information: Each system includes a slide box and a removable tray.
Delivery information: Index cards numbered from one to 100 for recording slide titles are supplied with each box.
Product Family Options
Product Information
  • Colour
    Material
    No. of slides
    W×D×H
    Availability
    Price
  • Blue
    PS
    100
    82×245×82 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 181,00 €
    181,00 €
    1 * 10 ST
       181,00 €
  • Green
    PS
    100
    82×245×82 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 181,00 €
    181,00 €
    1 * 10 ST
       181,00 €
  • Pink
    PS
    100
    82×245×82 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 181,00 €
    181,00 €
    1 * 10 ST
       181,00 €
  • White
    PS
    100
    82×245×82 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 181,00 €
    181,00 €
    1 * 10 ST
       181,00 €
  • Yellow
    PS
    100
    82×245×82 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 181,00 €
    181,00 €
    1 * 10 ST
       181,00 €