ContactUsLinkComponent
VWR®, Rack for Slide Boxes
VWR®, Rack for Slide Boxes
Catalog # 631-0742
Supplier:  VWR Collection
undefined
VWR®, Rack for Slide Boxes
Catalog # 631-0742
Supplier:  VWR Collection
Specifications
  • No. of slides:
    50/100
  • Description:
    Stackable drawer
  • Material:
    Acrylic
  • W×D×H:
    227×264×118 mm
  • Pk:
    1
Specifications
About this item
Storage racks for microscopy slides make organisation of samples easy.
  • Clear acrylic allows the labels on the slide boxes to be seen
  • Boxes can be placed in rows or columns
  • Sorted by colour to create an easy coding system
Non skid rubber feet allow for secure bench top placement, whilst pre-drilled holes enable under the counter or wall mounting.