Orders LinkContactUsLinkComponent
Slide boxes
Slide boxes
Supplier:   KARTELL
Slide boxes
Supplier:  KARTELL
Certificates
Slide boxes
3 Options Available Below
Product Details & Documents
High impact resistant slide boxes with removable lids.
  • Well spaced slots for easy handling of slides
  • Numbering for cross reference with corresponding index card
  • Locating lugs so boxes can be stacked
W×D×H: 83×98×38 mm (for 25); 97×230×35 mm (for 50); 280×230×35 mm (for 100)
Product Family Options
Product Information
  • Colour
    Material
    No. of slides
    W×D×H
    Availability
    Price
  • Grey
    PS
    25
    83×98×38 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 21,70 €
    21,70 €
    1 * 1 ST
       21,70 €
  • Grey
    PS
    50
    97×230×35 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,10 €
    28,10 €
    1 * 1 ST
       28,10 €
  • Grey
    PS
    100
    280×230×35 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 39,10 €
    39,10 €
    1 * 1 ST
       39,10 €