Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Storage cryobox, Nalgene®

Storage cryobox, Nalgene®
    
 
Storage cryobox, Nalgene®
Storage cryobox, Nalgene®

 

 
 
 

PC, white.

  • Suitable for mechanical freezer or vapor phase liquid nitrogen storage
  • Vial type : Cryobank tubes

Box with transparent lid for storage of various sizes of vials, bottles and bags in storage areas, refrigerators and mechanical freezers (–196 to +121 °C).

Double your cryobank storage capacity and cut storage costs by 50%. The Thermo Scientific™ Dense Storage Options use standard CryoBoxes™ with 13×13 tube separators and fit 169 1 ml tubes in a single CryoBox™. These standard-sized boxes are compatible with existing standard-sized lab freezer racks.

 

 

  • Catalog No:
  • 479-3321
  • 479-2505
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)

 

Product Family Options

 

  • Description
    For
    Colour
    W×D×H ext.
     
     
  • Without divider
    Cryobank
    White
    133×133×52 mm
       
       
    1 * 4 ST
    1 * 4 ST - 48,00 €
    48,00 €
     

      30/06/2026

       30/06/2026
    1 * 4 ST
       48,00 €

     
      Low Manufacturing Impact
     
    Specifications
    Cat. No.
    479-3321
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • With 13×13 divider, for 169 cryobank tubes (not included)
    Cryobank
    White
    133×133×52 mm
       
       
    1 * 4 ST
    1 * 4 ST - 50,00 €
    50,00 €
     

      30/06/2026

       30/06/2026
    1 * 4 ST
       50,00 €

     
      Low Manufacturing Impact
     
    Specifications
    Cat. No.
    479-2505
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact