Orders LinkContactUsLinkComponent
Cryoboxes, Nalgene®
Cryoboxes, Nalgene®
   Sustainable Options Available
Cryoboxes, Nalgene®
   Sustainable Options Available
Certificates
Cryoboxes, Nalgene®
3 Options Available Below
Limited Time Sale
Select items below are currently on sale
Product Details & Documents
White PC, autoclavable.
  • Durable, all-plastic CryoBoxes™ for short- or long-term storage
  • Plastic boxes are autoclavable for decontamination
  • Printed grid on plastic lids for easy inventory management
  • Chipboard/cardboard boxes have dividers and vents in base for good drainage and air circulation
  • Available with dividers from 25 to 169 tube storage
For ultra-low temperature storage of cryovials from –196 to +121 °C.
Transparent lid with printed grid permits viewing of contents and is keyed to prevent misalignment.
Accepts writing with cryo-markers.
Meet the needs of any budget with plastic or cardboard CryoBoxes™ designed to store cryogenic vials and tubes. Choose from a variety of durable plastic Thermo Scientific™ CryoBox™ styles with keyed lids and printed location grids, or select economical cardboard/chipboard CryoBoxes™ for short-term storage. The plastic boxes are optimised to fit our own brand of Nalgene™ and Nunc™ vials and microcentrifuge tubes best.
Specifications for Product Family
Specifications
  • Catalog No:
  • 479-3311
  • 479-5131
  • 479-0011
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
Specifications
Product Family Options
Product Information
  • Compartments
    For
    Colour
    W×D×H ext.
    Availability
    Price
  • 479-3311
    Nalgene® polycarbonate cryobox, weight: 0,52 kg
    Nalgene® polycarbonate cryobox, weight: 0,52 kg
    5×5
    Cryogenic vials 1,2 and 2,0 ml
    White
    76×76×52 mm
    Log in for availability
    Log in for availability
    1 * 8 ST
    1 * 8 ST - 62,00 €
    62,00 €
    Sale
    Valid until 30/06/2026
    SaleValid until 30/06/2026
    1 * 8 ST
       62,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    479-3311
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 479-5131
    Nalgene® polycarbonate cryobox, weight : 4 kg
    Nalgene® polycarbonate cryobox, weight : 4 kg
    9×9
    Cryogenic vials 1,2 and 2,0 ml
    White
    133×133×52 mm
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 49,00 €
    49,00 €
    Sale
    Valid until 30/06/2026
    SaleValid until 30/06/2026
    1 * 4 ST
       49,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    479-5131
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact
  • 479-0011
    Nalgene® polycarbonate cryobox, weight : 0,83 kg
    Nalgene® polycarbonate cryobox, weight : 0,83 kg
    9×9
    Cryogenic vials 5,0 ml
    White
    133×133×95 mm
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 62,00 €
    62,00 €
    Sale
    Valid until 30/06/2026
    SaleValid until 30/06/2026
    1 * 4 ST
       62,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    479-0011
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
      Low Manufacturing Impact