Orders LinkContactUsLinkComponent
Bottles, narrow neck, round, with thread
Bottles, narrow neck, round, with thread
Supplier:   GRAVIS
Bottles, narrow neck, round, with thread
Supplier:  GRAVIS
Certificates
Bottles, narrow neck, round, with thread
Bottles, narrow neck, round, with thread
5 Options Available Below
Product Details & Documents
In soda-lime glass.
  • White (215-2583, 215-0572 and 215-2584)
  • Brown (215-2585 and 215-2586)
These screwed round mini bottles are available in three volumes.
Ordering information: The capsules are ordered separately.
Documents
Product Family Options
Bottles
Product Information
  • Capacity
    Thread
    Ext.Ø
    Height
    Colour
    Availability
    Price
  • 2,5 ml
    13
    19 mm
    29 mm
    Clear
    Log in for availability
    Log in for availability
    1 * 360 ST
    1 * 360 ST - 278,00 €
    278,00 €
    1 * 360 ST
       278,00 €
  • 10 ml
    18 mm
    24,1 mm
    50,2 mm
    Clear
    Log in for availability
    Log in for availability
    1 * 195 ST
    1 * 195 ST - 138,00 €
    138,00 €
    1 * 195 ST
       138,00 €
  • 10 ml
    18 mm
    24,1 mm
    50,2 mm
    Brown
    Log in for availability
    Log in for availability
    1 * 195 ST
    1 * 195 ST - 138,00 €
    138,00 €
    1 * 195 ST
       138,00 €
  • 15 ml
    18 mm
    26,6 mm
    57,2 mm
    Clear
    Log in for availability
    Log in for availability
    1 * 216 ST
    1 * 216 ST - 160,00 €
    160,00 €
    1 * 216 ST
       160,00 €
  • 15 ml
    18 mm
    26,6 mm
    57,2 mm
    Brown
    Log in for availability
    Log in for availability
    1 * 216 ST
    1 * 216 ST - 158,00 €
    158,00 €
    1 * 216 ST
       158,00 €
Screw caps, Bakelite
Product Information
  • Catalog #
    Colour
    Thread
    Description
    Availability
    Price
  • Black
    18 mm
    with black Butyl/PTFE seal
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 133,00 €
    133,00 €
    1 * 100 ST
       133,00 €
Product Information
  • Catalog #
    Colour
    Thread
    Description
    Availability
    Price
  • Black
    18 mm
    with POLYLEX joint PH 18
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 29,10 €
    29,10 €
    1 * 100 ST
       29,10 €
Product Information
  • Catalog #
    Colour
    Thread
    Description
    Availability
    Price
  • Black
    18 mm
    with rubber liner
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 48,10 €
    48,10 €
    1 * 100 ST
       48,10 €
Screw caps, PP
Product Information
  • Catalog #
    Colour
    Description
    Thread
    Availability
    Price
  • Black
    with TRISEAL seal
    18 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 23,00 €
    23,00 €
    1 * 100 ST
       23,00 €
Product Information
  • Catalog #
    Colour
    Description
    Thread
    Availability
    Price
  • White
    with TRISEAL seal
    13 mm
    Log in for availability
    Log in for availability
    1 * 360 ST
    1 * 360 ST - 113,00 €
    113,00 €
    1 * 360 ST
       113,00 €
Complementary Products
Product Information
  • Catalog #
    Description
    Thread
    Colour
    Availability
    Price
  • 215-2681
    with black Butyl/PTFE seal
    18 mm
    Black
    Log in for availability
    1 * 100 ST - 133,00 €
  • 216-0573
    with POLYLEX joint PH 18
    18 mm
    Black
    Log in for availability
    1 * 100 ST - 29,10 €
  • 215-2696
    with rubber liner
    18 mm
    Black
    Log in for availability
    1 * 100 ST - 48,10 €