Packaging bottles, narrow neck, round, HDPE, with screw caps, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
HDPE, translucent or amber with PP screw cap.
- Good chemical resistance
- Heat resistant from –100 to +120 °C
- Leakproof
Economical, robust bottles for storing and transporting powders, liquids or pastes. Amber bottles to protect light-sensitive substances.
Caution: The product(s) on this page are outside the scope of the EU Directive 2019/904.
Specifications
- Catalog No:
- 215-1432
- 215-1007
- 215-1415
- 215-1414
- 215-1430
- 215-1416
- 215-1431
- 215-1017
- 215-1411
- 215-1410
- 215-1009
- 215-1412
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement