Culture media bottles, square, sterile, Nalgene®
PETG.
- Leakproof closure, break-resistant and durable
- Transparency and reduced permeability to carbon dioxide and oxygen allow media storage up to 6 months
- Bottles and closures are radiation sterilised and non-pyrogenic
An inexpensive alternative to glass bottles for storage and transport of media. Available with or without a natural HDPE screw cap closure. The caps for the 30 to 1000 ml bottles are a natural off-white colour. The cap for the 2000 ml bottle is white. Bottles and closures are not autoclavable. Packaged in shrink-wrapped trays.
- Catalog No:
- 216-0382
- 216-0309
- 216-0308
- 216-0306
- 216-0384
- 216-0383
- 216-0609
- 216-0312
- 216-0311
- 216-0381
- 216-0379
- 216-0380
- 216-0307
- 216-0310
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Frequently Bought Together