Jerricans, Nalgene®
HDPE, translucent with white PP screw cap.
- Good chemical resistance
- Temperature resistant from −100 to +120 °C
- Stable sturdy design with integral graduations in gallons and litres, leakproof closure
Good stability through a large base and low centre of gravity. The spout is long enough for accurate pouring of liquids. With tethered sealing cap that cannot be lost. Recessed base offers additional support during pouring.
HDPE, translucent with white PP screw cap.
- Good chemical resistance
- Temperature resistant from −100 to +120 °C
- Stable sturdy design with integral graduations in gallons and litres, leakproof closure
Good stability through a large base and low centre of gravity. The spout is long enough for accurate pouring of liquids. With tethered sealing cap that cannot be lost. Recessed base offers additional support during pouring.
- Catalog No:
- 216-8691
- 216-8692
- 216-8693
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
Product Family Options
- CapacityW×D×HThread
Low Manufacturing Impact Specifications
Cat. No.216-8691Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact Low Manufacturing Impact Specifications
Cat. No.216-8692Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact Low Manufacturing Impact Specifications
Cat. No.216-8693Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationNAR Certificate of RetirementLow Manufacturing Impact