Orders LinkContactUsLinkComponent
Bubble counters
Bubble counters
100 mm
Catalog # 201-9100
Supplier:   Lenz Laborglas
Bubble counters
Catalog # 201-9100
Supplier:  Lenz Laborglas
Bubble counters
Bubble counters
100 mm
Price
37,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
For monitoring gas flow in distillation apparatus.
Documents
Specifications
  • Height:
    100 mm
  • Cat. No.:
    201-9100