Orders LinkContactUsLinkComponent
Dropping bottles, Lucas
Dropping bottles, Lucas
Supplier:   MUELLER KREMPEL
Dropping bottles, Lucas
Supplier:  MUELLER KREMPEL
Certificates
Dropping bottles, Lucas
Dropping bottles, Lucas
7 Options Available Below
Product Details & Documents
Amber glass, without closure.
  • Good chemical and physical properties
  • Use in applications that do not involve high thermal stresses
  • Please order screw caps and drop dispensers separately
Brown coloured to protect light-sensitive materials.
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • 5 ml
    Log in for availability
    Log in for availability
    1 * 310 ST
    1 * 310 ST - 173,00 €
    173,00 €
    1 * 310 ST
       173,00 €
  • 10 ml
    Log in for availability
    Log in for availability
    1 * 187 ST
    1 * 187 ST - 105,00 €
    105,00 €
    1 * 187 ST
       105,00 €
  • 15 ml
    Log in for availability
    Log in for availability
    1 * 152 ST
    1 * 152 ST - 84,70 €
    84,70 €
    1 * 152 ST
       84,70 €
  • 20 ml
    Log in for availability
    Log in for availability
    1 * 180 ST
    1 * 180 ST - 101,00 €
    101,00 €
    1 * 180 ST
       101,00 €
  • 30 ml
    Log in for availability
    Log in for availability
    1 * 111 ST
    1 * 111 ST - 66,20 €
    66,20 €
    1 * 111 ST
       66,20 €
  • 50 ml
    Log in for availability
    Log in for availability
    1 * 105 ST
    1 * 105 ST - 68,90 €
    68,90 €
    1 * 105 ST
       68,90 €
  • 100 ml
    Log in for availability
    Log in for availability
    1 * 68 ST
    1 * 68 ST - 52,80 €
    52,80 €
    1 * 68 ST
       52,80 €
Drip closures for Lucas dropper bottles
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Vertical dropper for medium viscosity oils; Tamper evident closure with dropper insert for openings according to DIN-18, for medium viscosity oils
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 33,50 €
    33,50 €
    1 * 100 ST
       33,50 €
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Vertical dropper for aqueous solutions; White tamper evident closure with dropper insert for openings according to DIN-18 for low surface tension aqueous solutions; also containing ethanol up to 15%
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 33,50 €
    33,50 €
    1 * 100 ST
       33,50 €
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Vertical dropper for thinly flowing, essential oils; Tamper evident closure with dropper insert for openings according to DIN-18
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 33,50 €
    33,50 €
    1 * 100 ST
       33,50 €