Orders LinkContactUsLinkComponent
Dissolved oxygen bottles
Dissolved oxygen bottles
Supplier:   HIRSCHMANN
Dissolved oxygen bottles
Supplier:  HIRSCHMANN
Certificates
Dissolved oxygen bottles
Dissolved oxygen bottles
2 Options Available Below
Product Details & Documents
Oxygen bottles according to Winkler, with white inscription label.
  • The adjusted capacity is individually marked on the bottle
  • A unique serial number is engraved on the bottle and stopper
Product Family Options
Product Information
  • Capacity
    NS stopper
    Availability
    Price
  • 612-2794
    Oxygen bottles acc. to Winkler
    Oxygen bottles acc. to Winkler
    100-150 ml
    14/23
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,10 €
    36,10 €
    1 * 1 ST
       36,10 €
  • 215-1245
    Oxygen bottles acc. to Winkler
    Oxygen bottles acc. to Winkler
    200-300 ml
    19/26
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 40,40 €
    40,40 €
    1 * 1 ST
       40,40 €