ContactUsLinkComponent
Centrifuge bottles, PC, with screw cap, Nalgene®
Centrifuge bottles, PC, with screw cap, Nalgene®
Catalog # 525-2471
   Low Manufacturing Impact
undefined
Centrifuge bottles, PC, with screw cap, Nalgene®
Catalog # 525-2471
   Low Manufacturing Impact
Specifications
  • Ø×L:
    61,9×145,8 mm
  • Capacity:
    250 ml
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    4
Specifications
About this item
PC, round bottom, autoclavable with PP screw closure.
  • Transparent, excellent chemical resistance
  • Withstands forces up to 27 500 ×g
  • Use with adapter 525-2401