ContactUsLinkComponent
Replacement stopcock for aspirator bottles, Azlon®
Replacement stopcock for aspirator bottles, Azlon®
Catalog # 215-3164
Supplier:  SciLabware
Replacement stopcock for aspirator bottles, Azlon®
Catalog # 215-3164
Supplier:  SciLabware
Specifications
  • Description:
    Replacement stopcock for octagonal aspirator bottles
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
PP, heavy-duty.
  • Very good chemical resistance
  • Leakproof
Boss-fitting stopcock is suitable for carboys/aspirator bottles 215-3161 to 215-3163.